Semimicro Spatula with One Tapered End, One Rounded End
Semimicro Spatula with One Tapered End, One Rounded End
Actual product may vary
Semimicro Spatula with One Tapered End, One Rounded End
Fisherbrand Semimicro Spatula are designed for transferring small quantities from semimicro centrifuge and test tubes
Have Questions?
Catalog NumberLength (Metric)
FIS14-37418 cm
Catalog number FIS14-374
Price (SGD)
19.00
EA
Add to cart
Length (Metric):
18 cm
Price (SGD)
19.00
EA
Add to cart
  • Our spatulas are designed to exceed performance expectations -This is the Fisherbrand Advantage
  • Available in variety of lengths
  • Stainless-steel construction
  • Tip thickness: 0.035 in. (0.89mm)
  • One end tapered to 0.14 in. (3.5mm) rounded tip for use in conical-bottom tubes
  • Rounded end has 0.25 in. (6.6mm) rounded tip for regular tubes
Specifications
Product TypeSemiSpatula, Micro
For Use With (Equipment)Semi-micro centrifuge and conical bottom and regular test tubes
For Use With (Application)Designed to transfer small quantities from semimicro centrifuge and test tubes
Length (English)7 in.
IncludesDouble ended with tapered to 0.14 in. rounded tip at one end and rounded with 0.25 in. rounded tip on the other end
MaterialStainless Steel
Length (Metric)18 cm
Unit SizeEA