Chemi-Scraper™ Spatulas
Chemi-Scraper™ Spatulas
Actual product may vary
Chemi-Scraper™ Spatulas
Fisherbrand Chemi-Scraper™ Spatulas are treated for corrosion resistance and are designed for a non-sticking surface.
Have Questions?
Catalog NumberLength (Metric)
FIS14-37318 cm
Catalog number FIS14-373
Price (SGD)
25.00
EA
Add to cart
Length (Metric):
18 cm
Price (SGD)
25.00
EA
Add to cart
Our spatulas are designed to exceed performance expectations -This is the Fisherbrand Advantage.
  • Type 410 stainless-steel with bright satin finish
  • Stem dia.: 0.16 in. (4mm)
  • One rounded and one square end, each approx. 2L x 0.31 in.W (51 x 7.9mm)
Specifications
Product TypeSpatula, Chemi-Scraper™
FinishBright Satin
Length (English)7 in.
IncludesDouble ended with 2 L x 0.31 in. W rounded on one end and squared on other end
MaterialStainless Steel (410)
Length (Metric)18 cm
Unit SizeEA