▸
Histology and Cytology
▸
Tissue Fixatives
▸
Safetex™ Cytology Fixative
Actual product may vary
Andwin Scientific
Safetex™ Cytology Fixative
Have Questions?
Catalog Number
Product Type
FIS14-372-46
Fixative
Catalog number
FIS14-372-46
Price (SGD)
129.00
Pack
Add to cart
Product Type:
Fixative
Price (SGD)
129.00
Pack
Add to cart
Specifications
Product Type
Fixative
Quantity
29.6mL
Formulation
Water and Alcohol soluble Polyethylene Glycol
For Use With (Application)
Industrial use
Form
Liquid