Scoopula™ Spatula
Scoopula™ Spatula
Actual product may vary
Fisherbrand
Scoopula™ Spatula
Dig out materials, core, and stir easily
Have Questions?
Catalog NumberLength (Metric)
FIS14-357Q15 cm
Catalog number FIS14-357Q
Price (SGD)
-
Length (Metric):
15 cm
Dig out materials, core, and stir easily
  • Minimizes spilling in handling small amounts of material
  • More versatile
  • One pointed end, one blunt end
  • Stainless-steel
  • Does not rust
Specifications
ShapePointed End, Blunt End
Product TypeScoopula™
For Use With (Application)Minimizes spilling in handling small amounts of material, dig out materials, core and stir easily
Length (English)6 in.
IncludesOne pointed end and one blunt end
MaterialStainless Steel
Length (Metric)15 cm
Unit SizePK