Hard Rubber Spatulas
Hard Rubber Spatulas
Actual product may vary
John M. Maris Company
Hard Rubber Spatulas
Nonsparking, wide, flexible blade
Have Questions?
Catalog NumberLength (Metric)Length (Metric) Blade
FIS14-355B26 cm15.2 cm
Catalog number FIS14-355B
Price (SGD)
-
Length (Metric):
26 cm
Length (Metric) Blade:
15.2 cm
Nonsparking, wide, flexible blade
  • Integral handle
  • Can be used with materials that affect metals adversely
  • Washable
  • Sterilizable
Specifications
SterilitySterile
Product TypeSpatula
For Use With (Application)Designed to used with materials that affect Metals adversely, for mixing, weighing, cleaning, blending and many other uses
Length (English) Blade6 in.
Length (English)10.25 in.
Length (Metric) Blade15.2 cm
Width (English) Blade1.14 in.
Width (Metric) Blade2.9 cm
MaterialHard Rubber
Handle TypeIntegral
Length (Metric)26 cm
Unit SizeEA