Weighing Funnels
Weighing Funnels
Actual product may vary
Weighing Funnels
Weighing dish and sample transfer funnel in one.
Have Questions?
Catalog NumberDiameter (Metric)Top
FIS14-353D25 mm
Catalog number FIS14-353D
Price (SGD)
250.00
PK
Add to cart
Diameter (Metric)Top:
25 mm
Price (SGD)
250.00
PK
Add to cart
Weighing dish and sample transfer funnel in one.
  • Borosilicate glass funnels with tapered pouring necks aid microtransfer of solids and liquids
  • No rocking or moving while weighing—flat bottom keeps funnel steady while on balance
  • Use in temperatures to 454°C (849°F)
Assortment pack of five includes one each of the following (diameter x L): 10 x 35mm, 15 x 40mm, 20 x 45mm, 25 x 55mm and 32 x 68mm.
Specifications
Diameter (Metric)Top25 mm
Diameter (Metric) Stem10.25mm (OD); 8.12mm (ID)
MaterialBorosilicate Glass
Height (Metric)55 mm
For Use With (Application)Used for microtransfer of solids and liquids
Max. Temperature (English)849°F
Max. Temperature (Metric)454°C
Product TypeWeighing Funnel
Unit SizePK