Additional Mesh Inserts
Additional Mesh Inserts
Actual product may vary
Bel-Art
Additional Mesh Inserts
For use with Mini-Sieve™ micro sieve set.
Have Questions?
Catalog NumberScreen Size
FIS14-306BSet of 8 (25 Mesh, 35 Mesh, 45 Mesh, 60 Mesh, 80 Mesh, 120 Mesh, 170 Mesh, 230 Mesh)
Catalog number FIS14-306B
Price (SGD)
596.00
PK
Add to cart
Screen Size:
Set of 8 (25 Mesh, 35 Mesh, 45 Mesh, 60 Mesh, 80 Mesh, 120 Mesh, 170 Mesh, 230 Mesh)
Price (SGD)
596.00
PK
Add to cart
  • Made from phosphor-bronze
  • Removable
  • Includes pack of 8 phosphor-bronze screen inserts, 1 each of 25, 35, 45, 60, 80, 120, 170 and 230 standard mesh
Specifications
DescriptionRemovable
TypeSieve Insert
Diameter (English) Outer2.2 in.
For Use With (Equipment)Mini-Sieve micro sieve set
IncludesPack of 8 phosphor-bronze screen inserts, 1 each of 25, 35, 45, 60, 80, 120, 170 and 230 standard mesh
Diameter (Metric) Outer56 mm
Frame MaterialPlastic
Screen SizeSet of 8 (25 Mesh, 35 Mesh, 45 Mesh, 60 Mesh, 80 Mesh, 120 Mesh, 170 Mesh, 230 Mesh)
Mesh MaterialPhosphor Bronze
Unit SizePK