Fines Collector for Model L3P Sonic Sifter Separators
Actual product may vary
Advantech
Fines Collector for Model L3P Sonic Sifter Separators
For use with L3P sonic sifter separators
Have Questions?
Catalog NumberFor Use With (Equipment)
FIS14-294-20L3P sonic sifter separators
Catalog number FIS14-294-20
Price (SGD)
252.00
Each
For Use With (Equipment):
L3P sonic sifter separators
Price (SGD)
252.00
Each
For use with L3P sonic sifter separators
  • Fines Collector stops dust at bottom of air column in sifter and transmits oscillary motion to air column
Specifications
Product TypeFines Collector
For Use With (Application)Stops dust at bottom of sifter air column, transmits oscillary motion to air column
For Use With (Equipment)L3P sonic sifter separators
Unit SizeEach