▸
Shakers
▸
Shaker Accessories
▸
Diaphragm for Model L3P Sonic Sifter Separators
Actual product may vary
Advantech
Diaphragm for Model L3P Sonic Sifter Separators
For use with L3P sonic sifter separators
Have Questions?
Catalog Number
For Use With (Equipment)
FIS14-294-17
L3P sonic sifter separators
Catalog number
FIS14-294-17
Price (SGD)
18.00
Each
Add to cart
For Use With (Equipment):
L3P sonic sifter separators
Price (SGD)
18.00
Each
Add to cart
For use with L3P sonic sifter separators
Dust Stop/Diaphragm transmits oscillary motion to air column and keeps particles within column
Specifications
Product Type
Diaphragm
For Use With (Equipment)
L3P sonic sifter separators