▸
Tubing
▸
Rubber and Silicone Tubing
▸
Bioprene™ Continuous Biopharm Tubing
Actual product may vary
Watson-Marlow
Bioprene™ Continuous Biopharm Tubing
Suitable for upstream and downstream biopharmaceutical processes.
Have Questions?
Change view
Catalog Number
Tubing Size
Thickness (English)
FIS14-285-001
Bore: 1.6 mm (1/16 in.)
1/16 in.
FIS14-285-003
Bore: 3.2 mm (1/8 in.)
1/16 in.
2 Options
Catalog number
FIS14-285-001
Price (SGD)
1,087.00
Pack
Add to cart
Tubing Size:
Bore: 1.6 mm (1/16 in.)
Thickness (English):
1/16 in.
Price (SGD)
1,087.00
Pack
Add to cart
Offers exceptionally long peristaltic life, ensuring process security
Broad chemical compatibility; especially resistant to strong acids, alkalis and oxidizing agents
Widely applicable in biopharmaceutical manufacturing operations such as metering, transfer and filtration
Ideal for media feed and pH control in fermentation and metering tablet coating materials
Suitable for a variety of applications in the food and beverage industry, especially product transfer and metering
Operating pressures of 0-30 psi can be achieved
LoadSure tube element (P/N 14-285-009) increases the pressure capability up to a maximum of 100 psi
Specifications
Overall Length (Metric)
15 m
Thickness (English)
1/16 in.
Thickness (Metric)
1.6 mm
Min. Operating Temperature (Metric)
5°C
Tubing Size
Bore: 1.6 mm (1/16 in.)
Color
Opaque Beige
Max. Operating Temperature (Metric)
80°C
Product Type
Tubing