Heavy-Duty Scoops
Heavy-Duty Scoops
Actual product may vary
Heavy-Duty Scoops
Sturdy HDPE material.
Have Questions?
Catalog NumberCapacity (Metric)Size
FIS14-2431.89 LSmall
Catalog number FIS14-243
Price (SGD)
160.00
EA
Add to cart
Capacity (Metric):
1.89 L
Size:
Small
Price (SGD)
160.00
EA
Add to cart
  • Large Scoops are fabricated, heavy duty polyethylene scoop
  • With many industrial applications
  • Large scoops are sturdy and available in two sizes
Specifications
Product TypeHeavy Duty Scoop
MaterialHDPE
Capacity (English)0.5 gal.
Capacity (Metric)1.89 L
SizeSmall
ColorWhite
For Use With (Application)Designed for many industrial applications
Dimensions (L x W x H)5.87 x 6 x 13.5 in. (14.9 x 15.24 x 34.29 cm)
Height (English)13.5 in.
Height (Metric)34.29 cm
Length (English)5.87 in.
Length (English) Handle5.5 in.
Length (Metric)14.9 cm
Length (Metric) Handle13.97 cm
Width (English)6 in.
Width (Metric)15.24 cm
Unit SizeEA