Sterileware™ Sampler Scoops
Actual product may vary
Fisherbrand
Sterileware™ Sampler Scoops
Polystyrene plastic sampler scoops are designed for retrieving samples from a larger source.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (English)Quantity
FIS14-241-10C8 oz.100/Box
FIS14-241-10A2 oz.100/Box
FIS14-241-10B4 oz.100/Box
FKS117665184 oz.100/Box
Catalog number FIS14-241-10C
Price (SGD)
859.00
Case
Capacity (English):
8 oz.
Quantity:
100/Box
Price (SGD)
859.00
Case
Manufactured from polystyrene plastic. SP Scienceware™ Sterileware™ Sampler Scoops are designed for retrieving samples from a larger source.
  • For use with wide-mouth containers
  • One-piece, molded of high-impact polystyrene plastic
  • Remain upright on flat surfaces
  • Can be used as weighing boat
  • Made from FDA grade materials
  • Individually sealed and sterilized
  • For one time use and disposal
  • Capacity and volumes are estimated, not meant for taking exact sample weights or volumes
  • Stated volumes represent typical heaping samples
  • Lot stamped for accurate lot tracing
  • Includes a Certificate of Processing for sterilization
Specifications
Quantity100/Box
SterilitySterile
AutoclavableNot Autoclavable
Capacity (English)8 oz.
Capacity (Metric)250 mL
ColorWhite
Certifications/ComplianceFDA
DisposableSingle-use
Product TypeScoop
For Use With (Equipment)Wide mouth containers
For Use With (Application)Perfect shape and size for taking samples, especially from wide-mouth containers
Length (English)7.5 in.
Size13.5 x 5.4 x 4.9 cm (LxWxH) Bowl
IncludesCertificate of processing for sterilization
MaterialPolystyrene
Handle TypeExtra long
Length (Metric)19.1 cm
Unit SizeCase