SP Scienceware™ Reusable Polypropylene Scoops
Actual product may vary
Bel-Art
SP Scienceware™ Reusable Polypropylene Scoops
Designed for retrieving samples from a larger source. SP Scienceware™ Reusable Polypropylene Scoops are a valuable multi-purpose lab tool.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Length (Metric) HandleDimensions (L x W)
FIS14-237-51100 mL10.3 cm32.7 x 12.5 x 9.4 cm
FIS14-237-3250 mL5.6 cm19.1 x 5.4 x 4.9 cm
FIS03-410-80360 mL12.7 cm20.5 x 4.1 x 2.9 cm
FIS14-237-6125 mL12.7 cm22.1 x 5.1 x 3.3 cm
FIS14-237-122 mL3.2 cm9.4 x 3.2 x 2.2 cm
FIS14-237-275 mL4.8 cm14.1 x 4.4 x 2.7 cm
Catalog number FIS14-237-5
Price (SGD)
70.00
Special offer
Ends: 06-Sep-2026
100.00
Save 30.00 (30%)
Pack
Capacity (Metric):
1100 mL
Length (Metric) Handle:
10.3 cm
Dimensions (L x W):
32.7 x 12.5 x 9.4 cm
Price (SGD)
70.00
Special offer
Ends: 06-Sep-2026
100.00
Save 30.00 (30%)
Pack
Designed for retrieving samples from a larger source. SP Scienceware™ Reusable Polypropylene Scoops are a valuable multi-purpose lab tool.
  • Rigid form scoop
  • Flat bottom keeps scoops upright when filled or empty
  • Can be used as weighing boat
  • Manufactured from polypropylene
  • Steam autoclavable at 121°C (250°F)
  • Complies with FDA regulations for food contact
  • Capacity/volumes are estimated and are not meant for taking exact sample weights/volumes
  • Stated volumes represent typical heaping samples
Specifications
AutoclavableAutoclavable
Capacity (English)37.195 oz.
Capacity (Metric)1100 mL
ColorTranslucent
Certifications/ComplianceFDA
DisposableReusable
Product TypeScoop
FinishSmooth
For Use With (Application)Designed to remain upright when filled or empty allowing them to be used as weighing boats or to be placed on a flat surface for ease sample collection and dispensing
Length (English)12.87 in.
Width (English)4.94 in.
Width (Metric)12.5 cm
Dimensions (L x W)32.7 x 12.5 x 9.4 cm
Length (English) Handle4.06 in.
Length (Metric) Handle10.3 cm
MaterialPolypropylene
Length (Metric)32.7 cm
Unit SizePack