Aerosol Filter Tips, Sterile, Wide Bore
Actual product may vary
Axygen
Aerosol Filter Tips, Sterile, Wide Bore
Prevent cross-contamination with robotic barrier tips. Axygen™ Aerosol Filter Tips, Sterile are e-beam irradiated to prevent product degradation, comply with USP 3127 (Heavy Metal Contamination) and are free of leachable additives. Available in a wide variety of tip styles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
FIS14-222-7801 to 200 μL
FIS14-222-768100 to 1000 μL
Catalog number FIS14-222-780
Price (SGD)
1,264.00
Case
Volume (Metric):
1 to 200 μL
Price (SGD)
1,264.00
Case
  • Technologically advanced filter material allows free passage of air while blocking aerosol contamination.
  • Filter membrane contains no leachable additives.
  • Axygen™ Maxymum Recovery™ surface improves accuracy and ensures maximum sample recovery.
  • RNase-/DNase-free
  • Nonpyrogenic
  • Human gDNA- and PCR-inhibition-free
Specifications
Volume (Metric)1 to 200 μL
FilteredFiltered
SterilitySterile
ColorClear
Certifications/ComplianceUSP 3127
Clean ClaimsDNase, RNase, and Pyrogen Free
FormatRack
Tip StyleGenomic/Wide Orifice
Packaging96 Tips/Rack, 10 Racks/Pk, 5 Packs/Cs
For Use With (Equipment)Gilson Pipetman P200