Aerosol Filter Tips, Sterile
Actual product may vary
Axygen
Aerosol Filter Tips, Sterile
Prevent cross-contamination with robotic barrier tips. Axygen™ Aerosol Filter Tips, Sterile are e-beam irradiated to prevent product degradation and are free of leachable additives. Available in a wide variety of tip styles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Tip StyleFormat
FIS14-222-7771 to 200 μLMaxymum RecoveryRack
FIS14-222-7810.5 to 20 μLMaxymum RecoveryRack
Catalog number FIS14-222-777
Price (SGD)
282.00
Pack
Volume (Metric):
1 to 200 μL
Tip Style:
Maxymum Recovery
Format:
Rack
Price (SGD)
282.00
Pack
  • Technologically advanced filter barrier pipette tips
  • Ideal for DNA amplification and microbiological applications where trace contamination can invalidate results
  • Filter allows free passage of air while preventing cross-contamination by blocking the passage of aerosols or liquids
  • Nonpyrogenic, clear polyethylene filters are free from DNA inhibitors and leachable additives
  • Every filter is inserted into the tip robotically to ensure freedom from nuclease contamination
  • Packaged in sealed-bottom racks with lids to prevent contamination
  • Wide variety of tip styles available in a range of packaging quantities:
    • Maxymum Recovery™ technology provides excellent accuracy and prevents wastage of precious samples
    • Wide bore tips are ideal for pipetting viscous materials, genomic DNA or delicate cell cultures
Specifications
Volume (Metric)1 to 200 μL
Volume (Metric) Dispensing1 to 200 μL
FilteredFiltered
SterilitySterile
ColorClear
Rack ColorClear
AutoclavableAutoclavable
Certifications/ComplianceISO 13485
Clean ClaimsDNase, RNase, and Pyrogen Free
For Use With (Application)DNA Amplification, Microbiological Applications
FormatRack
Tip StyleMaxymum Recovery
PackagingRack
Quantity960/Pk.
Product TypeUniversal Fit Filter Pipet Tip
Includes12 Racks x 96 Tips
Green FeaturesNo Intentionally Added PFAS
ACT LabelNo
For Use With (Equipment)Gilson Pipetman P200