10μL Microvolume Tips
Actual product may vary
Axygen
10μL Microvolume Tips
Axygen Universal Tips designed to fit a wide variety of single and multi-channel pipettes, compatible with most popular brands of pipettes. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
Have Questions?
Catalog NumberVolume (Metric)
FIS14-222-7370.5 to 10 μL
Catalog number FIS14-222-737
Price (SGD)
70.00
Pack
Volume (Metric):
0.5 to 10 μL
Price (SGD)
70.00
Pack
Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. Nonpyrogenic
  • Microvolume
Specifications
Volume (Metric)0.5 to 10 μL
FilteredNon-filtered
SterilityNon-sterile
ColorClear
AutoclavableAutoclavable
ModelT-300
Certifications/ComplianceUSP 3127
Clean ClaimsDNase, Rnase Free
For Use With (Application)Allows recovery of costly samples with improved precision and accuracy
Tip StyleMicro Point
Packaging1,000 Tips/Pk., 20 Packs/Cs.
Quantity1000 Tips/Pk., 20 Pk./Cs.
Product TypePipette Tips
IncludesTips, Pack, Case
For Use With (Equipment)Gilson Pipetman P10, Gilson Pipetman P2