1000 μL Universal Pipetter Tips: Wide Bore
Actual product may vary
Axygen
1000 μL Universal Pipetter Tips: Wide Bore
Compatible with most popular brands of pipettes
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSterilityFormat
FIS14-222-703SterileRack
FIS14-222-699Non-sterileBulk
Catalog number FIS14-222-703
Price (SGD)
1,070.00
Case
Sterility:
Sterile
Format:
Rack
Price (SGD)
1,070.00
Case
Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are nonpyrogenic and RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
(This is the suggested replacement for T-1005-WB-C-L)
  • These polypropylene tips are nonpyrogenic and RNase-/DNase-free.
  • Nonpyrogenic
  • Wide-Bore
Specifications
Volume (Metric)100 to 1000 μL
FilteredNon-filtered
SterilitySterile
ColorClear
AutoclavableAutoclavable
Clean ClaimsDNase, RNase, DNA, and PCR inhibitor Free
FormatRack
Tip StyleGenomic/Wide Orifice, Universal
Packaging100 Tips/Rack, 10 Racks/Pk, 5 Packs/Cs
For Use With (Equipment)Universal Fit