1000 μL Universal Pipetter Tips: Bevelled
Actual product may vary
Axygen
1000 μL Universal Pipetter Tips: Bevelled
Compatible with most popular brands of pipettes
Have Questions?
Catalog NumberSterilityColorFormat
FIS14-222-690Non-sterileBlueBulk
Catalog number FIS14-222-690
Price (SGD)
80.00
Pack
Sterility:
Non-sterile
Color:
Blue
Format:
Bulk
Price (SGD)
80.00
Pack
Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Beveled
Specifications
Volume (Metric)100 to 1000 μL
FilteredNon-filtered
SterilityNon-sterile
ColorBlue
AutoclavableAutoclavable
ModelT-1000-B
Certifications/ComplianceEN ISO 13485 compliant, BSE/TSE compliant, Non-Pyrogenic, USP Class VI tested, Human DNA Free, DNase/RNase Free, Quality Control Tested
Clean ClaimsDNase, RNase, and Pyrogen free
FormatBulk
Tip StyleBeveled, Universal
Packaging1,000 Tips/Pk., 5 Packs/Cs.
Quantity1000 Tips/Pk., 5 Pk./Cs.
Product TypePipette Tips
For Use With (Equipment)Universal Fit