PCR Tubes with Flat Cap, 0.2 mL
Actual product may vary
Axygen
PCR Tubes with Flat Cap, 0.2 mL
Designed to fit standard 96-well blocks. Axygen™ PCR Tubes with Flat Cap, 0.2m are constructed of non-sterile polypropylene. Each conical tube features thin and consistent wall thickness. Tight-fitting cap secures contents and prevents evaporation. Available in an assortment of colors.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorProduct Type
FIS14-222-260AssortedThin Wall PCR Tube
FIS14-222-262ClearThin Wall PCR Tube
FKS11376044ClearThin Wall PCR Tube
FIS14-222-283WhiteMaxymum Recovery PCR Tube
Catalog number FIS14-222-260
Price (SGD)
180.00
Pack
Color:
Assorted
Product Type:
Thin Wall PCR Tube
Price (SGD)
180.00
Pack
Designed to fit standard 96-well blocks. Axygen™ PCR Tubes with Flat Cap, 0.2m are constructed of non-sterile polypropylene. Each conical tube features thin and consistent wall thickness. Tight-fitting cap secures contents and prevents evaporation. Available in an assortment of colors.
  • Quantitative PCR compatible tubes for a wide range of real-time PCR instruments
  • Ultra-thin and consistent wall thickness allows precise thermal transfer
  • Even insertion cap seals full length for a tight fit that prevents sample evaporation
  • Maxymum Recovery™ option provides ultra-low retention and high yield
  • Certified RNase-/DNase-free and nonpyrogenic
  • Manufactured with 99.9% pure virgin polypropylene
  • Available in a wide variety of colors to support color-coding
Specifications
Certifications/ComplianceRNase, DNase, pyrogen-free
ColorAssorted
Volume (Metric)0.2 mL
Quantity1000/Pk., 10Pk./Cs.
Closure MaterialPolypropylene
For Use With (Equipment)Standard 96-well blocks of real-time PCR instruments and thermal transfer applications
ShapeConical
SterilityNon-sterile
MaterialPolypropylene
Closure TypeFlat Cap
FormatTube
Product TypeThin Wall PCR Tube
Unit SizePack