8-Strip PCR Tubes, 0.2 mL
Actual product may vary
Axygen
8-Strip PCR Tubes, 0.2 mL
Available in a variety of colors for easy sample identification. Axygen™ 8-Strip PCR Tubes, 0.2mL are constructed of sturdy polypropylene. Each non-sterile, conical tube features thin and consistent wall thickness. Sold with flat or domed caps to prevent sample loss or without caps.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorClosure TypeQuantity
FIS14-222-252ClearFlat Cap125/Pk., 1250/Cs.
FIS14-222-251ClearDomed Cap125/Pk., 1250/Cs.
FIS14-222-248AssortedSold Separately125/Pk., 1250/Cs.
FIS14-222-253GreenSold Separately125/Pk., 1250/Cs.
FIS14-222-257YellowSold Separately125/Pk., 1250/Cs.
Catalog number FIS14-222-252
Price (SGD)
319.00
Pack
Color:
Clear
Closure Type:
Flat Cap
Quantity:
125/Pk., 1250/Cs.
Price (SGD)
319.00
Pack
Available in a variety of colors for easy sample identification. Axygen™ 8-Strip PCR Tubes, 0.2mL are constructed of sturdy polypropylene. Each non-sterile, conical tube features thin and consistent wall thickness. Sold with flat or domed caps to pr
  • PCR tubes strips designed to fit a wide variety of thermal cyclers
  • Ultra-thin and consistent wall thickness allows precise thermal transfer
  • Domed or flat insertion caps ensure a tight fit that prevents sample evaporation
  • Certified RNase-/DNase-free and nonpyrogenic
  • Manufactured with 99.9% pure virgin polypropylene
  • Available in a variety of colors for easy sample identification
Specifications
Certifications/ComplianceRNase, DNase, pyrogen-free
ColorClear
Volume (Metric)0.2 mL
Quantity125/Pk., 1250/Cs.
Closure MaterialPolypropylene
For Use With (Equipment)Thermal cyclers for thermal transfer applications
ShapeConical
SterilityNon-sterile
MaterialPolypropylene
Max. Force4000 x g
Closure TypeFlat Cap
FormatStrip
Product TypeThin Wall 8-Strip PCR Tube
Unit SizePack