MaxyClear Snaplock Microtubes, 2.0 mL
Actual product may vary
Axygen
MaxyClear Snaplock Microtubes, 2.0 mL
Ensure an efficient seal that stays closed during centrifugation. Axygen™ MaxyClear Snaplock Microtubes, 2.0mL provide excellent visibility and feature a piercing port for syringe access. Available in a wide variety of colors. Maxymum Recovery™ and sterile options available.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorSterilityPackaging
FIS14-222-183ClearNon-sterile250 Tubes/Pk., 10 Packs/Cs.
FIS14-222-180ClearNon-sterile500 Tubes/Pk., 10 Packs/Cs.
FIS14-222-181ClearSterile250 Tubes/Pk., 10 Packs/Cs.
Catalog number FIS14-222-183
Price (SGD)
52.00
Pack
Color:
Clear
Sterility:
Non-sterile
Packaging:
250 Tubes/Pk., 10 Packs/Cs.
Price (SGD)
52.00
Pack
Axygen MaxyClear microcentrifuge tubes are available in a variety of colors. Axygen Maxymum Recovery™ surface and sterile options are also available. SnapLock microcentrifuge tubes have frosted cap surfaces and frosted side panels, ideal for labeling. Each cap features a piercing port in the center of the cap. Microcentrifuge tubes with no caps are available for some sizes.
  • 0.6 mL, 1.5 mL, and 2.0 mL clear microcentrifuge tubes are boil proof. Colored tubes are not boil-proof.
  • Co-polymer tubes offer exceptional strength and chemical resistance. Not to be autocalvable or boil-proof.
  • Clear microcentrifuge tubes are boil-proof. Colored tubes are not boil-proof.
  • Maximum G-force is 14,000 RCF.
  • RNase-/DNase-free and nonpyrogenic
  • Sterility Assurance Level (SAL) of 10−6
  • Color - Assorted: Each pack contains a mixture of blue, green, violet, orange, red, and yellow.
  • Clear microcentrifuge tubes are boil-proof. Colored tubes are not boil-proof.
  • Color - Spectrum: 2 packs each of blue, green, orange, red, and yellow.
Specifications
Cap StyleSnap Cap
Certifications/ComplianceRNase, DNase, pyrogen-free
Closure MaterialPolypropylene
AutoclavableAutoclavable
Capacity (Metric)2 mL
ColorClear
MaterialHomopolymer
Increments0.25 mL
No. per Pack250/Pk.
Packaging250 Tubes/Pk., 10 Packs/Cs.
SterilityNon-sterile
Product LineMaxymum Recovery
GraduatedYes
Closure TypeSnap Cap
Max. RCF20,000 x g
Graduations0.1 to 2 mL
Product TypeLocking Snap-cap Microtube
Unit SizePack