Standard Write-On Sample Bags
Actual product may vary
Nasco
Standard Write-On Sample Bags
White, write-on strip makes sample identification easier and more convenient.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W)Capacity (English)
FIS14-209-30050.8 x 25.4 cm (20 x 10 in.)123 oz.
FIS01-812-13038 x 25.4 cm (15 x 10 in.)92 oz.
FKS1173179638 x 25.4 cm (15 x 10 in.)92 oz.
FKS1174179650.8 x 25.4 cm (20 x 10 in.)123 oz.
FIS01-812-12750.8 x 38 cm (20 x 15 in.)184 oz.
FKS1175179650.8 x 38 cm (20 x 15 in.)184 oz.
Catalog number FIS14-209-300
Price (SGD)
754.00
Pack
Dimensions (L x W):
50.8 x 25.4 cm (20 x 10 in.)
Capacity (English):
123 oz.
Price (SGD)
754.00
Pack
White, write-on strip makes sample identification easier and more convenient.
  • Closing tab has one round and one flat wire to ensure that bag is closed securely
  • Fold tab over tightly 4 to 5 times when closing
  • Volume measurements are approximate
  • White, write-on strip makes sample identification easier and more convenient
  • Paper-like finish eliminates ink smearing and provides surface for any writing instrument
  • Sterilized
  • Note: Should not be used at temperatures above 82°C
  • Bags can be frozen to any temperature, even liquid nitrogen
Specifications
AutoclavableNot Autoclavable
Closure TypeClosing Tab
ColorClear
DisposableSingle-use
For Use With (Application)Accommodates large size samples and eliminate reducing into smaller pieces
IncludesWhite, Write-on Strip
MaterialPolyethylene
SterilitySterile
Fill Volume123 oz.
Latex FreeYes
No. per Pack250/Pk.
Outer PackagingCardboard
Protective Inner PackagingPlastic
Quantity250/Pk.
Sterilization MethodEthylene Oxide Gas
Temperature Range (English)-320°F to +180°F
Temperature Range (Metric)-196°C to +82°C
Thickness (English)4 mil
Dimensions (L x W)50.8 x 25.4 cm (20 x 10 in.)
Length (English)20 in.
Width (English)10 in.
Product TypeGiant-size Sample Bag
Capacity (English)123 oz.
Capacity (Metric)3637 mL
Length (Metric)50.8 cm
Width (Metric)25.4 cm
Thickness (Metric)0.102 mm
Unit SizePack