Lab Matting
Lab Matting
Actual product may vary
Lab Matting
Lab Matting
Lab Matting
Easy to clean, easy to cut.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W)Length (English)Length (Metric)
FIS14-203B100 ft. x 30 in. (30 m x 76 cm)100 ft.30.4 m
FIS14-203A15 ft. x 30 in. (4.4 m x 76 cm)15 ft.4.57 m
Catalog number FIS14-203B
Price (SGD)
571.00
EA
Add to cart
Dimensions (L x W):
100 ft. x 30 in. (30 m x 76 cm)
Length (English):
100 ft.
Length (Metric):
30.4 m
Price (SGD)
571.00
EA
Add to cart
Easy to clean, easy to cut.
Protects drawers, shelves, and benches from chemicals, spills, and dirt.
  • Clear polyethylene ribbed matting cushions instruments, reduces glassware breakage
  • Five times thicker (0.036 in.) than normal shelf liner, but won't ripple; lies flat
Specifications
Product TypeLab Mat
MaterialPolyethylene
Length (English)100 ft.
Length (Metric)30.4 m
Width (English)30 in.
Width (Metric)762 mm
Dimensions (L x W)100 ft. x 30 in. (30 m x 76 cm)
ColorClear
For Use With (Application)Protects drawers, shelves, and benches from chemicals, spills, and dirt
Thickness9 mm (0.036 in.)
Quantity1 Ea.
Unit SizeEA