Pure Natural Rubber Tubing in 12ft. Lengths
Pure Natural Rubber Tubing in 12ft. Lengths
Actual product may vary
Pure Natural Rubber Tubing in 12ft. Lengths
Contains dry natural rubber.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (English) InnerDiameter (English) OuterColor
FIS14-178D5/16 in.0.437 in.Amber
FIS14-178A0.156 in.1/4 in.Amber
FIS14-178B0.188 in.0.312 in.Amber
FIS14-150-5C1/4 in.3/8 in.Black
Catalog number FIS14-178D
Price (SGD)
40.00
EA
Add to cart
Diameter (English) Inner:
5/16 in.
Diameter (English) Outer:
0.437 in.
Color:
Amber
Price (SGD)
40.00
EA
Add to cart
Contains dry natural rubber
  • Made with dry natural rubber
  • Available in variety of sizes
  • Offered in Black and Amber
Specifications
Certifications/ComplianceZZ-T-831D Types I and III, U.S. Pharmacopeia Class VI
AutoclavableAutoclavable
Diameter (English) Inner5/16 in.
Diameter (Metric) Inner7.9 mm
MaterialRubber
Diameter (English) Outer0.437 in.
Diameter (Metric) Outer11.1 mm
Overall Length (English)12 ft.
Overall Length (Metric)3.7 m
Thickness (English)0.0625 in.
Thickness (Metric)1.6 mm
ColorAmber
Product TypeTubing
Unit SizeEA