Pharma-50 Tubing
Actual product may vary
Corning
Pharma-50 Tubing
Translucent, pharmaceutical-grade silicone tubing for ultrapure fluid processing.
Have Questions?
Catalog NumberDiameter (English) InnerDiameter (English) OuterThickness (English)
FIS14-177-1073/8 in.5/8 in.0.125 in.
Catalog number FIS14-177-107
Price (SGD)
735.00
Each
Diameter (English) Inner:
3/8 in.
Diameter (English) Outer:
5/8 in.
Thickness (English):
0.125 in.
Price (SGD)
735.00
Each
Designed for applications that require the transfer of high-purity fluids where contamination is a concern, particularly pharmaceutical and biotechnological manufacturing processes. Made from tear-resistant silicone platinum-cured elastomer.
  • Hydrophobic surface
  • Highly flexible and resilient
  • Low extractables; no imparted taste or odor
  • Contains no organic plasticizers, phthalates, latex additives, peroxide by-products, chlorophenyls, or PCBs
  • Stable over a wide temperature range
  • Sterilize by autoclave, EtO, or gamma radiation
  • Durometer hardness: Shore A, 50
Not intended for hospital or patient care use, or for use as an implant.
Specifications
Certifications/ComplianceExceeds USP Class VI Plastic Test requirements and meets EP monograph “Silicone Elastomer for Closures and Tubing”. Manufactured according to Bulk Pharmaceutical Chemicals GMPs at an FDA-registered, ISO 9002-certified healthcare facility.
AutoclavableAutoclavable
Diameter (English) Inner3/8 in.
Diameter (Metric) Inner9.5 mm
MaterialPlatinum-cured Silicone
Diameter (English) Outer5/8 in.
Diameter (Metric) Outer15.8 mm
Overall Length (English)50 ft.
Thickness (English)0.125 in.
Thickness (Metric)3.2 mm
For Use With (Application)For ultrapure liquid transfer
Maximum Pressure14 psi
Unit SizeEach