Tygon™ ND 100-80 Tubing
Actual product may vary
Saint-Gobain
Tygon™ ND 100-80 Tubing
Tygon™ ND 100-80 Tubing for medical device manufacturers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (English) InnerDiameter (English) OuterThickness (English)
FIS14-171-2840.02 in.0.06 in.0.02 in.
FIS14-171-2820.01 in.0.03 in.0.01 in.
Catalog number FIS14-171-284
Price (SGD)
-
Diameter (English) Inner:
0.02 in.
Diameter (English) Outer:
0.06 in.
Thickness (English):
0.02 in.
Specifically formulated in a higher durameter that enables high resistance to kinking and collapse
Tygon™ ND 100-80 tubing is made from a biocompatible polymer that is not formulated with DEHP. This tubing is crystal clear for easy visualization of fluid flow. The physical properties of Tygon ND 100-80 enable manufacturing of smaller diameter tubes with a high resistance to kinking and collapse.
  • High rigidity for kink resistance
  • Compatible with blood
  • Crystal clear for easy visualization of fluid flow
  • Glass-smooth inner bore
  • Reduced flexibility for suction applications
  • Tygon™ ND 100-80 has met the requirements of Biological Test for Plastics Class VI, as described in USP <88> (2017)
  • Lot-to-lot consistency for reproducible results
  • Non-toxic and non-contaminating
  • Smooth, polished inner wall
  • Slips easily over fittings and grips securely for simple lab set-ups
  • Contains no BPA or phthalates
  • Standard sizes available to hold full vacuum at room temperature
Specifications
Diameter (English) Inner0.02 in.
Diameter (Metric) Inner0.51 mm
MaterialTygon ND-100-80
Diameter (English) Outer0.06 in.
Diameter (Metric) Outer1.52 mm
Thickness (English)0.02 in.
Thickness (Metric)0.51 mm
For Use With (Application)Medical grade
Port SizeNeedle Gauge: 23
ColorClear
Product TypeTubing
Unit SizePack