Check Order Status
General-Purpose Rubber Tubing in 12ft. (3.7m) Lengths
Actual product may vary
General-Purpose Rubber Tubing in 12ft. (3.7m) Lengths
Seamless rubber tubing with nonslip surface and flexibility
Have Questions?
Catalog Number
Diameter (Metric) Inner
Diameter (Metric) Outer
Thickness (Metric)
Color
FIS14-158D
6.35 mm
10.9 mm
2.36 mm
Black
Catalog number
FIS14-158D
Price (SGD)
43.00
EA
Add to cart
Diameter (Metric) Inner:
6.35 mm
Diameter (Metric) Outer:
10.9 mm
Thickness (Metric):
2.36 mm
Color:
Black
Price (SGD)
43.00
EA
Add to cart
Made with dry natural rubber
Available in variety of sizes
Offered in Black and Red
Specifications
Diameter (English) Inner
1/4 in.
Diameter (Metric) Inner
6.35 mm
Material
Rubber
Diameter (English) Outer
0.43 in.
Diameter (Metric) Outer
10.9 mm
Overall Length (English)
12 ft.
Overall Length (Metric)
3.7 m
Thickness (English)
0.093 in.
Thickness (Metric)
2.36 mm
For Use With (Application)
General purpose
Color
Black