Check Order Status
Solid Neoprene Rubber Stoppers
Actual product may vary
Solid Neoprene Rubber Stoppers
Recommended where rubber would be attacked or deteriorated
Have Questions?
Change view
Catalog Number
Size
Stopper Number
FIS14-141J
7
7
FIS14-141L
9
9
2 Options
Catalog number
FIS14-141J
Price (SGD)
95.00
PK
Add to cart
Size:
7
Stopper Number:
7
Price (SGD)
95.00
PK
Add to cart
Recommended where rubber would be attacked or deteriorated
Solid neoprene
Resists petroleum products, fats, oils and organic chemicals
Specifications
For Use With (Application)
Resists petroleum products, fats, oils and organic chemicals
Length (Metric)
2.5 cm
Size
7
Color
Green
Diameter (Metric) Bottom
3 cm
Diameter (Metric) Top
3.7 cm
Material
Neoprene
Product Type
Solid Stopper
Stopper Number
7