Solid Neoprene Rubber Stoppers
Solid Neoprene Rubber Stoppers
Actual product may vary
Solid Neoprene Rubber Stoppers
Have Questions?
Catalog NumberStopper Number
FIS14-141H6.5
Catalog number FIS14-141H
Price (SGD)
103.00
PK
Add to cart
Stopper Number:
6.5
Price (SGD)
103.00
PK
Add to cart
  • Solid neoprene
  • Resists petroleum products, fats, oils and organic chemicals
Specifications
For Use With (Application)Resists petroleum products, fats, oils and organic chemicals
Length (Metric)0.25 cm
Size6.5
ColorGreen
Diameter (Metric) Bottom2.7 cm
Diameter (Metric) Top3.4 cm
MaterialNeoprene
Product TypeSolid Stopper
Stopper Number6.5
Unit SizePK