Two-Hole Rubber Stoppers
Two-Hole Rubber Stoppers
Actual product may vary
Two-Hole Rubber Stoppers
Assorted sizes 00 to 15
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric) TopLength (Metric)SizeStopper Number
FIS14-140P6.4 cm2.5 cm1212
FIS14-140K4.1 cm2.5 cm88
FIS14-140M5 cm2.5 cm1010
Catalog number FIS14-140P
Price (SGD)
97.00
PK
Add to cart
Diameter (Metric) Top:
6.4 cm
Length (Metric):
2.5 cm
Size:
12
Stopper Number:
12
Price (SGD)
97.00
PK
Add to cart
Assorted sizes 00 to 15
  • Hole size: 3 to 5mm
Specifications
Diameter (Metric) Hole0.5 cm
Diameter (Metric) Inner5 mm
For Use With (Application)General purpose use
Length (Metric)2.5 cm
Size12
Total Positions2
ColorBlack
Diameter (Metric) Bottom5.4 cm
Diameter (Metric) Top6.4 cm
MaterialNatural Rubber
Product TypeTapered Stopper (2 holes)
Stopper Number12
Unit SizePK