One-Hole Rubber Stoppers
One-Hole Rubber Stoppers
Actual product may vary
One-Hole Rubber Stoppers
One hole rubber stopper
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSizeStopper Number
FIS14-135R1212
FIS14-135P1010
FIS14-135M88
FIS14-135B00
FIS14-135G55
FIS14-135D22
FIS14-135N99
FIS14-135A000
FIS14-135L77
FIS14-135E33
FIS14-135C11
Catalog number FIS14-135R
Price (SGD)
102.00
PK
Add to cart
Size:
12
Stopper Number:
12
Price (SGD)
102.00
PK
Add to cart
One hole rubber stopper
  • Available in Size No. 00 to 15
  • Hole size: 3 to 5mm
Specifications
Diameter (Metric) Hole0.5cm
Diameter (Metric) Inner5mm
For Use With (Application)Inexpensive black natural rubber/SBR blend for general purpose uses
Length (Metric)2.5 cm
Size12
Total Positions1
ColorBlack
Diameter (Metric) Bottom5.4 cm
Diameter (Metric) Top6.7 cm
MaterialNatural Rubber
Product TypeTapered Stopper (1 hole)
Stopper Number12
Unit SizePK