Stoppers made from soft, resilient, natural rubber with safe convenient countersunk holes, stoppers feature excellent aging characteristics with high resistance to deformation
Have Questions?
Catalog Number
Color
Stopper Number
FIS14-131D
Black
6
Catalog number FIS14-131D
Price (SGD)
123.00
PK
Add to cart
Color:
Black
Stopper Number:
6
Price (SGD)
123.00
PK
Add to cart
Simple twist-off seals provide convenient one, two, or three-hole use
Solid stoppers are vacuum-tested at 30 in. of mercury
High resistance to deformation—even after repeated autoclaving
Each stopper has its size number in raised figures on top
Specifications
For Use With (Application)Multi-purpose applications