Slide Holder
Actual product may vary
Jaece Industries
Slide Holder
Provides an inexpensive way for drying excess fluids from flat surfaces. Fisherbrand™ Slide Holder is constructed of chemical-resistant HDPE.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSlide Capacity
FIS14-127-50B78
FIS01-257-73315
Catalog number FIS14-127-50B
Price (SGD)
105.00
Each
Slide Capacity:
78
Price (SGD)
105.00
Each
Chemical-resistant high-density polyethylene holder
  • Convenient-to-use receptacle holds both 3 x 1 in. or 75 x 25 mm slides during evacuation and cleaning
  • Light, strong, chemical-resistant high-density polyethylene holder provides an inexpensive way for drying excess fluids from flat surfaces
  • 10° angle of slots facilitates clear viewing of desired slide
  • Beveled edges provide easy access to channels
Specifications
TypeSlide Holder
Slide Capacity78
AutoclavableNot Autoclavable
ColorNatural
For Use With (Equipment)Microscope Slide
Length (English) Slide3 in.
Length (Metric) Slide75 mm
MaterialHDPE
SterilityNon-sterile
Width (English) Slide1 in.
Width (Metric) Slide25 mm
Unit SizeEach