Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Stoppers and Closures
▸
Stoppers
▸
Identi-plug™ Plastic Foam Stoppers
Actual product may vary
Jaece Industries
Identi-plug™ Plastic Foam Stoppers
For test tubes and flasks
Have Questions?
Change view
Catalog Number
Diameter (Metric) Inner
Length (Metric)
FIS14-127-40F
5 cm
5.63 cm
FIS14-127-40D
3.4 cm
4.44 cm
FIS14-127-40B
2 cm
3.5 cm
3 Options
Catalog number
FIS14-127-40F
Price (SGD)
209.00
Pack
Add to cart
Diameter (Metric) Inner:
5 cm
Length (Metric):
5.63 cm
Price (SGD)
209.00
Pack
Add to cart
For test tubes and flasks
Easily compressed to form a snug-fitting stopper
When removed from container, they spring back to original shape
Permit air exchange and can be used over a wide range of temperatures; ideal for use with drosophilia vials
Withstand constant use and sterilization
Autoclavable and chemically resistant
Specifications
Diameter (Metric) Inner
5 cm
For Use With (Equipment)
For 4.6 to 5cm test tubes and flasks openings
Length (Metric)
5.63 cm
Quantity
80
Autoclavable
Autoclavable
Color
White
Material
Plastic Foam
Product Type
Plug Stopper
Sterility
Sterile