Product Image
Jaece Industries
Plastic Foam Stoppers
For test tubes and flasks
Have Questions?
Catalog NumberColor
FIS14-127-40EAssorted
Catalog number FIS14-127-40E
Price (SGD)
133.00
Pack
Color:
Assorted
Price (SGD)
133.00
Pack
For test tubes and flasks
  • Easily compressed to form a snug-fitting stopper
  • When removed from container, they spring back to original shape
  • Permit air exchange and can be used over a wide range of temperatures; ideal for use with drosophilia vials
  • Withstand constant use and sterilization
  • Autoclavable and chemically resistant
Specifications
Diameter (Metric) Inner4.5 cm
For Use With (Equipment)For 3.5 to 4.5cm test tubes and flasks openings
Length (Metric)4.44 cm
AutoclavableAutoclavable
ColorAssorted
MaterialPlastic
Product TypePlug Stopper