Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Stoppers and Closures
▸
Stoppers
▸
Foam Plugs for Vials
Actual product may vary
Jaece Industries
Foam Plugs for Vials
These foam plugs are an economical option for providing temporary closure for narrow vials and test tubes.
Have Questions?
Catalog Number
Color
FIS14-127-40C
White
Catalog number
FIS14-127-40C
Price (SGD)
242.00
Pack
Add to cart
Color:
White
Price (SGD)
242.00
Pack
Add to cart
These foam plugs are an economical option for providing temporary closure for narrow vials and test tubes.
Fits vials from 21 to 26mm in diamter
Provided in a pack of 400
Specifications
Diameter (Metric) Inner
2.6cm
For Use With (Equipment)
For 21 to 26mm in diameter vials
Length (Metric)
4.44 cm
Quantity
400/pk
Autoclavable
Autoclavable
Color
White
Material
Foam
Product Type
Stopper