Polypropylene Freezer Storage Boxes
Actual product may vary
Polypropylene Freezer Storage Boxes
Polypropylene Freezer Storage Boxes
Polypropylene Freezer Storage Boxes
100-place polypropylene storage boxes provide compact, safe storage for 0.5, 1.5 and 2.0 mL tubes and cryo vials. Unlike cardboard boxes they will not break down if exposed to moisture or heavy use.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
FIS14-100-CGreen
FIS14-100-ANatural
FIS14-100-BBlue
FIS14-100-DOrange
FIS14-100-EPurple
FIS14-100-FBlack
FIS14-100FBlack
FIS14-100-GAssorted
Catalog number FIS14-100-C
Price (SGD)
123.00
Pack
Color:
Green
Price (SGD)
123.00
Pack
100-place polypropylene storage boxes provide compact, safe storage for 0.5, 1.5 and 2.0 mL tubes and cryo vials. Unlike cardboard boxes they will not break down if exposed to moisture or heavy use.
  • Great for organization on the bench, in the freezer or during transport
  • Can be used at temperatures -80 to +121°C (112 to +249°F)
  • Five color choices let you visually sort and organize samples of different types
  • Hinged lids with snap tab open easily and close securely; clear translucent color always lets you see inside
  • Opaque black boxes are ideal for light sensitive samples
  • Each box position is labeled with an alphanumeric grid
  • No slide features molded in bottom and lid provide secure stacking
Specifications
Closure TypeHinged Lid
ColorGreen
MaterialPolypropylene
ShapeSquare
For Use With (Application)Provide compact and safe storage for 0.5, 1.5 and 2.0ml tubes and cryo vials
For Use With (Equipment)0.5, 1.5 and 2.0mL tubes and cryo vials
Holds100 (0.5, 1.5 and 2 mL Tubes and Cryo Vials)
IncludesAlphanumeric Grid Label, Clear Translucent Hinged Lids with Snap Tab
No. of Places100
Product TypeCryo Storage Box
Quantity5 Pack
Temperature Range (English)-112°F to 249.8°F
Temperature Range (Metric)-80° to 121°C
Dimensions (L x W x H)14.3 x 14.3 x 5.6 cm (5.7 x 5.7 x 2.2 in.)
Length (Metric)14.3 cm
Width (Metric)14.3 cm
Height (Metric)5.6 cm
Unit SizePack