Flowmi™ Cell Strainers for 1000μL Pipette Tips
Actual product may vary
Flowmi™ Cell Strainers for 1000μL Pipette Tips
Flowmi™ Cell Strainers for 1000μL Pipette Tips
Flowmi™ Cell Strainers for 1000μL Pipette Tips
Flowmi™ Cell Strainers for 1000μL Pipette Tips
Bel-Art
Flowmi™ Cell Strainers for 1000μL Pipette Tips
Provides fast and efficient filtering of small volume samples (up to 1000 μL) prior to FLOW or FACS analysis (patent pending). Available in two porosity sizes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMesh Size
FIS14-100-15040 μm
FIS03-421-22870 μm
Catalog number FIS14-100-150
Price (SGD)
309.00
Pack
Mesh Size:
40 μm
Price (SGD)
309.00
Pack
  • Provide fast and efficient filtering of small volume samples (up to 1000 μL) prior to FLOW or FACS analysis
  • Preserves volume when filtering small sample volumes
  • Removes cell aggregates and creates uniform single cell suspensions
  • Decreases the potential for clogging of FLOW or FACS instruments by effectively filtering cellular debris
  • Compact tray holds 50 Flowmi Cell Strainers ready for press and fit attachment to tip; features a sliding cover for easy, one-handed use
  • Packaged in a re-sealable bag to maintain sterility when used in laminar flow hood or biohazard hood
  • Sterile, 40 μm or 70 μm porosity
Specifications
Mesh MaterialPolyethylene
ColorWhite
For Use With (Application)Provide fast and efficient filtering of small volume samples (up to 1000μL) prior to FLOW or FACS™ analysis
IncludesCompact tray
Product TypeCell Strainer
SterilitySterile
For Use With (Equipment)Pipette Tips, 1000 μL, including Fisherbrand Sure-One, VWR, Axygen, Nichiryo, Eppendorf and more
Mesh Size40 μm
Unit SizePack