Extension Ring Clamps
Extension Ring Clamps
Actual product may vary
Extension Ring Clamps
Shank of extension ring clamps to support rods with clamp holder
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (English) InnerDiameter (English) OuterLength (English)
FIS14-055C3.9 in.4.5 in.9.8 in.
FIS14-055A1.4 in.2 in.8.6 in.
FIS14-055B2.9 in.3.5 in.9.8 in.
Catalog number FIS14-055C
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (English) Inner:
3.9 in.
Diameter (English) Outer:
4.5 in.
Length (English):
9.8 in.
Manufactured from steel wire, zinc-plated for corrosion resistance.
Specifications
MaterialPlated Steel Wire
Length (English)9.8 in.
Length (Metric)25 cm
Diameter (English) Inner3.9 in.
Diameter (English) Outer4.5 in.
Diameter (Metric) Inner10 cm
Diameter (Metric) Outer11.4 cm
Product TypeExtension Ring
For Use With (Application)Shank of extension ring clamps to support rods with clamp holder
Unit SizeEA