Cast-Iron Rings
Cast-Iron Rings
Actual product may vary
Cast-Iron Rings
Cast-iron construction prevents breakage under normal use
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (English) InnerDiameter (English) OuterDiameter (Metric) InnerDiameter (Metric) Outer
FIS14-050EQ5.25 in.6 in.13 cm15 cm
FIS14-050CQ3.31 in.4 in.8.4 cm10 cm
FIS14-050BQ2.37 in.3 in.6 cm7.6 cm
FIS14-050DQ4.25 in.5 in.11 cm13 cm
FIS14-050AQ1.43 in.2 in.3.6 cm5.1 cm
Catalog number FIS14-050EQ
Price (SGD)
0.00
EA
Add to cart
Diameter (English) Inner:
5.25 in.
Diameter (English) Outer:
6 in.
Diameter (Metric) Inner:
13 cm
Diameter (Metric) Outer:
15 cm
Price (SGD)
0.00
EA
Add to cart
Cast-iron construction prevents breakage under normal use
  • Sturdy 0.01 in. (7.9mm) setscrew in integral clamp locks ring tightly to support stands, lattices, etc.
  • Ring center is standard 4.5 in. (11cm) from support rod center
  • 2 in. (51mm) Size ring has lugs for holding crucibles
Specifications
MaterialCast Iron
Diameter (English) Inner5.25 in.
Diameter (English) Outer6 in.
Diameter (Metric) Inner13 cm
Diameter (Metric) Outer15 cm
Product TypeCast Iron Ring
ShapeRound
For Use With (Application)Prevents breakage under normal use
Quantity1 Ea.
Unit SizeEA