Portable Pipet-Aid™ XP Pipet Controller
Actual product may vary
Drummond
Portable Pipet-Aid™ XP Pipet Controller
Designed to minimize operator fatigue
Have Questions?
Change viewbuttonViewtableView
Catalog NumberModelVoltage
FIS1368107w/o Gravity drain230 V
FIS13-681-07w/o Gravity drain230 V
FIS13-681-15Ew/o Gravity drain110 V
Catalog number FIS1368107
Price (SGD)
1,370.00
Each
Model:
w/o Gravity drain
Voltage:
230 V
Price (SGD)
1,370.00
Each
Smaller, lightweight and specifically designed to minimize operator fatigue
  • Lightweight and compact
  • Accepts most pipets from 1 to 100mL
  • Three-speed settings (fast, medium, slow) provide more precise control in filler mode and dispenser modes
  • EZ Grip nosepiece with double membrane filter
  • Easy-access battery replacement
  • Removable stand
  • Holster-type wall bracket
  • Powered by a 9-volt (Ni-MH) rechargeable battery
Specifications
Battery TypeNiMH
Compatible TipsEZ Grip TC Filter Nosepiece
AutoclavableNot Autoclavable
For Use WithPipette
Modelw/o Gravity drain
TypeFiller/Dispenser
Volume (Metric)1 to 100 mL
For Use With (Application)For rapid filling of large pipets
For Use With (Equipment)1 to 100mL pipets
IncludesWall bracket, 4 replacement filters and 220V recharger
Weight (English)7.5 oz.
Weight (Metric)212.6 g