PowerMark; Single-Color Printer Ribbons
PowerMark; Single-Color Printer Ribbons
Actual product may vary
PowerMark; Single-Color Printer Ribbons
PowerMark; Single-Color Printer Ribbons are industrial-grade ribbons uniquely designed with memory cells that communicate information to the BBP™85 and PowerMark™ sign and label printers.
Have Questions?
Catalog NumberColorWidth (English)
FIS135-16Green8.8 in.
Catalog number FIS135-16
Price (SGD)
-
Color:
Green
Width (English):
8.8 in.
  • One-color ribbon for printing a single color onto a label
  • Tough, smear-proof ribbon produces long-lasting printed text that can withstand years in industrial areas and outdoor conditions
  • R10000 series ribbons are based on a resin formulation that offers a high performance image with strong solvent and scratch resistance
  • Ribbon cartridge eliminates calibration by automatically communicating information to the printer
Specifications
ColorGreen
FormatRibbon
Product TypePrinter Ribbon
SterilityNon-sterile
For Use With (Application)For outdoors and in industrial areas
For Use With (Equipment)BBP™85, PowerMark™ Printer
Quantity1 Ea.
Length (English)200 ft.
Length (Metric)60.9 m
Width (English)8.8 in.
Unit SizeEA