Chart Recorders for Ultra-Low Temperature Freezers
Actual product may vary
Thermo Scientific
Chart Recorders for Ultra-Low Temperature Freezers
Record seven days of temperature performance at temperatures to -150° C.
Have Questions?
Catalog NumberFor Use With (Equipment)TypedimVoltage
FIS13-994-002Forma 900, HERAfreeze HFU-B, Revco ExF/DxF, TSE/TSD SeriesInkless Chart Paper (50 Pk.)
Catalog number FIS13-994-002
Price (SGD)
512.00
Each
For Use With (Equipment):
Forma 900, HERAfreeze HFU-B, Revco ExF/DxF, TSE/TSD Series
Type:
Inkless Chart Paper (50 Pk.)
Price (SGD)
512.00
Each
Record seven days of temperature performance at temperatures to -150° C using Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers. Recorders use 6 in. (15cm) chart paper and are available as factory or field installed.
Features:
  • Seven-day recorder
  • Functions to -150°C
  • Field or factory installed
Ordering information:
  • Field-installed units must be installed by a qualified professional
Specifications
TypeInkless Chart Paper (50 Pk.)
Temperature Range-150°C
For Use With (Equipment)Forma 900, HERAfreeze HFU-B, Revco ExF/DxF, TSE/TSD Series
Static Holding Time7 Days
Monitoring OptionsRecorder
Unit SizeEach