Specifications
Product TypeHeat Sensitive Recorder Chart
Diameter (Metric)301.49 mm
Diameter (English)11.87 in.
For Use With (Application)Thermal chart allows recorder to print variety of programmable grid configurations and simultaneously record up to four distinct processes
For Use With (Equipment)Honeywell DR 4500 Truline
No. of Ranges0
ShapeCircular
Temperature Range (Metric)None
Time RangeNone
Unit SizePack