Pipette Tips, 2 to 200 μL
Actual product may vary
BrandTech
Pipette Tips, 2 to 200 μL
Designed for high-quality micro-pipettes and suitable for various applications. BRAND™ 2 to 200 μL pipette tips ensure precise and efficient pipetting. Tips are cleanroom-produced, free from contaminants, and environmentally friendly.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPackaging
FIS13-889-174TipBox
FIS13-889-143Bagged (1000 Pk.)
Catalog number FIS13-889-174
Price (SGD)
62.00
Pack
Packaging:
TipBox
Price (SGD)
62.00
Pack
  • Engineered to fit the Transferpette™ line of micropipettes
  • Compatibility tested with other micropipettors from various manufacturers
  • Manufactured and packaged under certified cleanroom conditions according to ISO 14644-1, Class 7 and 8
  • Made from high-quality, pure raw materials, free of DiHEMDA and oleamide
  • No use of mold release agents such as stearates and erucamide in the production process
  • Palletized BIO-CERT™ quality tips are guaranteed to be free from DNA, DNase, RNase, ATP, and endotoxins
  • Thin-walled for high translucency, easy ejection, and low waste
  • Graduations at 20 and 100 μL for quick volume checks
  • Available either in bulk packaging or convenient, ecological rack-and-stack formats
  • Bulk tips are yellow and placed in clear, resealable bags
  • Racked tips are colorless and placed into yellow-colored tip trays
  • TipBoxes are made of 100% PP and are fully recyclable
  • The TipRack refilling system reduces waste by more than 20% compared to the TipBox
Specifications
Volume (Metric)2 to 200 μL
FilteredNon-filtered
SterilityNon-sterile
ColorColorless
AutoclavableAutoclavable
Clean ClaimsDNase, RNase, ATP, and Endotoxin Free
FormatBox
Tip StyleUniversal
PackagingTipBox
Product TypePipette Tips
DisposableLimited-use
For Use With (Equipment)Universal Fit
GraduatedYes
MaterialPolypropylene
Graduations20, 100 μL
Unit SizePack