Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Cuvettes and Cells
▸
Cuvette Caps
▸
Cuvette Caps
Actual product may vary
BrandTech
Cuvette Caps
Provides tight seal, allows storage of samples to -20°C.
Have Questions?
Catalog Number
Color
FIS13-878-125
Blue
Catalog number
FIS13-878-125
Price (SGD)
116.00
Pack
Add to cart
Color:
Blue
Price (SGD)
116.00
Pack
Add to cart
Round cuvette caps
Fits ultra-micro cuvettes
Provide tight seal
Allows storage of samples to -20°
Specifications
Type
Cuvette Cap
Color
Blue
Disposable
Single-use
Material
Polyethylene
Quantity
100 Pack
For Use With (Equipment)
Ultramicro UV Cuvette
Shape
Round