Hemostats Kelly Forceps
Actual product may vary
Hemostats Kelly Forceps
Fisherbrand Hemostats Kelly Forceps are primarily used to control blood flow in surgical procedures. ASTM™ standard (F899-84) stainless steel.
Have Questions?
Catalog NumberTip Style
FIS13-812-14Curved
Catalog number FIS13-812-14
Price (SGD)
7.00
Each
Tip Style:
Curved
Price (SGD)
7.00
Each
Fisherbrand Hemostats Kelly Forceps are primarily used to control blood flow in surgical procedures. ASTM™ standard (F899-84) stainless steel.
  • Fisherbrand™ Hemostats Kelly Forceps are primarily used to control blood flow in large blood vessels and tubing during surgical procedures
  • Our forceps are designed to exceed perfromance expectations-This is the Fisherbrand Advantage.
  • 3-Stage Locking System – Variable blood flow control (Stage 0: complete flow to Stage 3: no flow)
  • Finger-ring handle designed for comfort, stability and ease of use
  • Multipurpose - Useful in educational, research or healthcare setting
  • Hand polished to mirror finish
  • Made from surgical stainless steel
  • Electronically passivated for corrosion resistance
  • Can be sterilized by Eto/gamma radiation
  • Rockwell hardness (HRC): 40-45
  • Autoclavable up to 135°C
  • Latex-free
Specifications
Product TypeHemostats Kelly Forcep
Certifications/ComplianceISO 9001, cCMP
MaterialStainless Steel
IncludesSerrated jaws, box lock joint
Tip StyleCurved
Length (English)5.5 in.
Length (Metric)14 cm
Unit SizeEach