Straight-Blade Operating Scissors
Actual product may vary
Straight-Blade Operating Scissors
Use these scissors for a variety of applications. Fisherbrand™ Straight-Blade Operating Scissors are forged from surgical-grade stainless steel.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Tip Style
FIS13-806-411.4 cmSharp/Sharp
FIS13-806-211.4 cmBlunt/Sharp
FIS13-808-414 cmSharp/Sharp
Catalog number FIS13-806-4
Price (SGD)
7.00
Each
Length (Metric):
11.4 cm
Tip Style:
Sharp/Sharp
Price (SGD)
7.00
Each
Use these scissors for a variety of applications. Fisherbrand™ Straight-Blade Operating Scissors are forged from surgical-grade stainless steel.
  • Fisherbrand™ Straight-Blade Operating Scissors.
  • Slope-cut and heavy beveled blades
  • Heavy body construction in a rounded English pattern
  • Electronically passivated
  • Autoclavable up to 135°
  • Surgical grade stainless steel
  • Can be sterilized by Eto/Gamma radiation
  • Rockwell Hardness (HRC): 48-50
  • Inspected per U.S
  • Government specifications for surgical instruments
  • Produced under ISO 9001, cGMP Guidelines
Specifications
Blade StyleStraight
Length (English)4.5 in.
Length (Metric)11.4 cm
AutoclavableAutoclavable
For Use With (Application)Surgical, Precision
Tip StyleSharp/Sharp
Unit SizeEach