Gauze Sponges
Actual product may vary
Gauze Sponges
Fluffy and soft
Have Questions?
Catalog NumberDimensions (L x W)ThicknessSterility
FIS13-761-524 x 4 in.12-PlyNon-sterile
Catalog number FIS13-761-52
Price (SGD)
21.00
Pack
Dimensions (L x W):
4 x 4 in.
Thickness:
12-Ply
Sterility:
Non-sterile
Price (SGD)
21.00
Pack
  • Made of 100% cotton with the cut edges folded in
  • Provide bulk, loft and high absorption for a wide range of prepping and dressing procedures
  • Sterile and nonsterile variety
Specifications
MaterialCotton
Product TypeGauze Sponge
SterilityNon-sterile
Length (English)4 in.
Length (Metric)10.16 cm
Width (Metric)10.16 cm
Width (English)4 in.
Quantity200 Pk.
Dimensions (L x W)4 x 4 in.
For Use With (Application)Ideal for wide range of prepping and dressing procedures
IncludesFolded-in cut edges
Thickness12-Ply
AbsorbancyHighly
Certifications/ComplianceUSP Type VII
Unit SizePack