PYREX™ Spot Plates
PYREX™ Spot Plates
Actual product may vary
PYREX™ Spot Plates
For observing color reactions with transmitted light
Have Questions?
Catalog NumberNo. of Wells
FIS13-748B9
Catalog number FIS13-748B
Price (SGD)
1,097.00
PK
Add to cart
No. of Wells:
9
Price (SGD)
1,097.00
PK
Add to cart
(This is the suggested replacement for 7223-34)
These pressed PYREX™ glass plates have 9 concave depressions (22mm O.D. x 7mm deep) which are useful for soil chemistry, color reactions, microchemical applications and microscopic observations.
Specifications
Depth (English)0.25 in.
Depth (Metric)7 mm
Diameter (English) Outer0.875 in.
Diameter (Metric) Outer22.2 mm
Dimensions (L x W)4 x 3.375 in. (100 x 7 mm)
For Use With (Application)Useful for soil chemistry, color reactions, microchemical applications, microscopic observations
Length (English)4 in.
Length (Metric)100 mm
No. of Wells9
Width (English)3.375 in.
Width (Metric)7 mm
MaterialBorosilicate Glass
TypeSpot Plate
Unit SizePK